ENA394768505 Enamine

5-carbamoylpyridine-3-carboxylic acid

  • MDL number MFCD18803436
  • Cas Number : 24242-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-44993
Availability details for
Quantity
  • Availability
  • Add to Cart