ENA394768507 Enamine

2-chloro-N-[4-(4-fluorophenyl)-5-methyl-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD07186521
  • Cas Number : 842954-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46065

Safety Information

msds https://www.enaminestore.com/catalog/EN300-46065
Availability details for
Quantity
  • Availability
  • Add to Cart