ENA394768515 Enamine

5-chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00068130
  • Cas Number : 128455-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 39...41
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55579

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55579
Availability details for
Quantity
  • Availability
  • Add to Cart