ENA394768521 Enamine

2-ethyl-2-(methylamino)butanenitrile

  • MDL number MFCD10694842
  • Cas Number : 55793-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59338

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59338
Availability details for
Quantity
  • Availability
  • Add to Cart