ENA394768522 Enamine

methyl 3-{[(tert-butoxy)carbonyl]amino}propanoate

  • MDL number MFCD12198407
  • Cas Number : 42116-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60129

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60129
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart