ENA394768524 Enamine

methyl 3-chloropropanoate

  • MDL number MFCD00039381
  • Cas Number : 6001-87-2
  • Molecular Weight : 122.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60618

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P271,P280,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-60618
pictograms GHS02,GHS05,GHS07
hstatements H226,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart