ENA394768530 Enamine

2-(3,4-difluorophenyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD12110826
  • Cas Number : 1156584-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64181

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64181
Availability details for
Quantity
  • Availability
  • Add to Cart