ENA394768531 Enamine

tert-butyl N-(cyanomethyl)-N-methylcarbamate

  • MDL number MFCD06658361
  • Cas Number : 180976-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-64595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64595
Availability details for
Quantity
  • Availability
  • Add to Cart