ENA394768555 Enamine

methyl 2-(1-methylpiperidin-4-ylidene)acetate

  • MDL number MFCD12756506
  • Cas Number : 154594-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77950

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77950
Availability details for
Quantity
  • Availability
  • Add to Cart