ENA394768556 Enamine

(1-benzofuran-6-yl)boronic acid

  • MDL number MFCD17017954
  • Cas Number : 851525-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129779

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-129779
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart