ENA394768584 Enamine

3H,4H-pyrido[3,4-d]pyrimidin-4-one

  • MDL number MFCD09999183
  • Cas Number : 19178-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92072
Availability details for
Quantity
  • Availability
  • Add to Cart