ENA394768594 Enamine

1,4-dioxaspiro[4.5]decan-7-amine

  • MDL number MFCD19373870
  • Cas Number : 49672-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129803
Availability details for
Quantity
  • Availability
  • Add to Cart