ENA394768595 Enamine

1-(2,2-diethoxyethyl)-2,4-difluorobenzene

  • MDL number MFCD11554294
  • Cas Number : 1443342-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96935

Safety Information

msds https://www.enaminestore.com/catalog/EN300-96935
Availability details for
Quantity
  • Availability
  • Add to Cart