ENA394768596 Enamine

1-(methoxymethyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD10686615
  • Cas Number : 67567-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-97340
Availability details for
Quantity
  • Availability
  • Add to Cart