ENA394768600 Enamine

2,4-difluoropyridine

  • MDL number MFCD04114162
  • Cas Number : 34941-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-99859
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart