ENA402266819 Enamine

4-(1-methyl-1H-1,2,4-triazol-3-yl)aniline

  • MDL number MFCD25371566
  • Cas Number : 1461708-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117919

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-117919
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart