ENA402266821 Enamine

2,2-difluoro-2-(4-methyl-1,3-thiazol-2-yl)acetic acid

  • MDL number MFCD25371568
  • Cas Number : 1461714-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122314
Availability details for
Quantity
  • Availability
  • Add to Cart