ENA402266830 Enamine

ethyl 2-[3-methyl-1-(nitromethyl)cyclopentyl]acetate

  • MDL number MFCD25371588
  • Cas Number : 1461706-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127918

Safety Information

msds https://www.enaminestore.com/catalog/EN300-127918
Availability details for
Quantity
  • Availability
  • Add to Cart