ENA402266831 Enamine

2-methyl-6-nitro-4-phenoxyquinoline

  • MDL number MFCD25371590
  • Cas Number : 1461706-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128284

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128284
Availability details for
Quantity
  • Availability
  • Add to Cart