ENA402266835 Enamine

benzyl[1-(2,3-difluorophenyl)-2,2,2-trifluoroethyl]amine

  • MDL number MFCD25371595
  • Cas Number : 1461706-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128826

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128826
Availability details for
Quantity
  • Availability
  • Add to Cart