ENA402266840 Enamine

2-{2-[4-(trimethylsilyl)-1H-1,2,3-triazol-1-yl]ethyl}-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD25371604
  • Cas Number : 1461706-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129507
Availability details for
Quantity
  • Availability
  • Add to Cart