ENA402266846 Enamine

N'-(1-cyclopropylethyl)(tert-butoxy)carbohydrazide

  • MDL number MFCD25371613
  • Cas Number : 1370600-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129814

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129814
Availability details for
Quantity
  • Availability
  • Add to Cart