ENA402266848 Enamine

tert-butyl N-[(1,1-dioxo-1lambda6-thian-2-yl)methyl]carbamate

  • MDL number MFCD25371615
  • Cas Number : 1461708-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129924
Availability details for
Quantity
  • Availability
  • Add to Cart