ENA402266850 Enamine

3-amino-5-tert-butyl-2,3-dihydro-1H-indol-2-one hydrochloride

  • MDL number MFCD25371617
  • Cas Number : 1461706-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130025
Availability details for
Quantity
  • Availability
  • Add to Cart