ENA402266851 Enamine

4-[(3-tert-butyl-1,2,4-oxadiazol-5-yl)(morpholin-4-yl)methyl]morpholine

  • MDL number MFCD25371618
  • Cas Number : 1461715-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-130032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130032
Availability details for
Quantity
  • Availability
  • Add to Cart