ENA402266859 Enamine

2-[(2-methoxyethyl)amino]propanoic acid hydrochloride

  • MDL number MFCD25371627

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130439

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130439
Availability details for
Quantity
  • Availability
  • Add to Cart