ENA402266867 Enamine

(4-amino-1-benzylpiperidin-4-yl)methanol dihydrochloride

  • MDL number MFCD25371638
  • Cas Number : 1461706-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130692
Availability details for
Quantity
  • Availability
  • Add to Cart