ENA402266868 Enamine

3-(benzylsulfanyl)-4-(methoxymethyl)benzoic acid

  • MDL number MFCD25371639
  • Cas Number : 1461707-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130695

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130695
Availability details for
Quantity
  • Availability
  • Add to Cart