ENA402266870 Enamine

1-(pyrrolidin-3-yl)-1H-1,3-benzodiazole dihydrochloride

  • MDL number MFCD25371641
  • Cas Number : 1461706-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130751

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130751
Availability details for
Quantity
  • Availability
  • Add to Cart