ENA402266871 Enamine

tert-butyl 3-[(2-nitrophenyl)amino]pyrrolidine-1-carboxylate

  • MDL number MFCD25371643
  • Cas Number : 1461707-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-130766

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130766
Availability details for
Quantity
  • Availability
  • Add to Cart