ENA402266873 Enamine

ethyl 4,4,4-trifluoro-2-methyl-3-(phenylimino)butanoate

  • MDL number MFCD25371645
  • Cas Number : 1461726-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130772

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130772
Availability details for
Quantity
  • Availability
  • Add to Cart