ENA402266875 Enamine

4-[4-(chloromethyl)-1-methyl-1H-pyrazol-3-yl]benzonitrile

  • MDL number MFCD25371648
  • Cas Number : 1461706-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130776

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130776
Availability details for
Quantity
  • Availability
  • Add to Cart