ENA402266879 Enamine

tert-butyl 1,3-dioxo-2-phenyl-octahydroimidazolidino[1,5-a]piperazine-7-carboxylate

  • MDL number MFCD25371653
  • Cas Number : 1461704-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130814

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130814
Availability details for
Quantity
  • Availability
  • Add to Cart