ENA402266882 Enamine

2-cyclopentyl-1-(pyridin-2-yl)ethan-1-one hydrochloride

  • MDL number MFCD25371659
  • Cas Number : 1461715-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130978

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130978
Availability details for
Quantity
  • Availability
  • Add to Cart