ENA402266884 Enamine

1-(2-fluorobenzoyl)pyrrolidin-3-amine hydrochloride

  • MDL number MFCD25371662
  • Cas Number : 1461705-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130983

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130983
Availability details for
Quantity
  • Availability
  • Add to Cart