ENA402266887 Enamine

5-nitro-2-(piperidine-1-carbonyl)pyridine

  • MDL number MFCD25371669
  • Cas Number : 1301214-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131072

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131072
Availability details for
Quantity
  • Availability
  • Add to Cart