ENA402266890 Enamine

2-cyclobutylmethanesulfonylethan-1-amine hydrochloride

  • MDL number MFCD25371672
  • Cas Number : 1461708-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131086
Availability details for
Quantity
  • Availability
  • Add to Cart