ENA402266891 Enamine

sodium (6-methoxy-1-oxo-2,3-dihydro-1H-inden-2-ylidene)methanolate

  • MDL number MFCD21863407
  • Cas Number : 1461727-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131090

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131090
Availability details for
Quantity
  • Availability
  • Add to Cart