ENA402266896 Enamine

methyl 2-{[(tert-butoxy)carbonyl]amino}-3-(furan-2-yl)propanoate

  • MDL number MFCD25371678
  • Cas Number : 1461704-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131114

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131114
Availability details for
Quantity
  • Availability
  • Add to Cart