ENA402266899 Enamine

tert-butyl 3-(5-methyl-1,2,4-oxadiazol-3-yl)morpholine-4-carboxylate

  • MDL number MFCD25371684
  • Cas Number : 1461714-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131151
Availability details for
Quantity
  • Availability
  • Add to Cart