ENA402266900 Enamine

1-[(4-aminonaphthalen-1-yl)sulfanyl]-N,N-dimethylformamide

  • MDL number MFCD25371685
  • Cas Number : 1461706-67-1
  • Molecular Weight : 246.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131155
Availability details for
Quantity
  • Availability
  • Add to Cart