ENA402266902 Enamine

N-methoxycyclopentanamine hydrochloride

  • MDL number MFCD25371687
  • Cas Number : 1461707-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131208
Availability details for
Quantity
  • Availability
  • Add to Cart