ENA402266904 Enamine

3-amino-5-(propan-2-yl)-2,3-dihydro-1H-indol-2-one hydrochloride

  • MDL number MFCD25371692
  • Cas Number : 1461705-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131243
Availability details for
Quantity
  • Availability
  • Add to Cart