ENA402266905 Enamine

7-fluoro-1-(propan-2-yl)-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD25371693
  • Cas Number : 1085542-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131244

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131244
Availability details for
Quantity
  • Availability
  • Add to Cart