ENA402266909 Enamine

4-bromo-2-[(4-methoxyphenyl)methoxy]-1-(trifluoromethyl)benzene

  • MDL number MFCD25371698
  • Cas Number : 1461705-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131317
Availability details for
Quantity
  • Availability
  • Add to Cart