ENA402266911 Enamine

ethyl 2-(chlorosulfonyl)-5-fluorobenzoate

  • MDL number MFCD25371700
  • Cas Number : 1461706-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-131327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131327
Availability details for
Quantity
  • Availability
  • Add to Cart