ENA402266912 Enamine

ethyl 7-bromo-3-methyl-1-benzofuran-2-carboxylate

  • MDL number MFCD25371702
  • Cas Number : 196799-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68...70
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131350

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131350
Availability details for
Quantity
  • Availability
  • Add to Cart