ENA402266915 Enamine

methyl 4-[4-(chloromethyl)-1,3-oxazol-2-yl]benzoate

  • MDL number MFCD25371705
  • Cas Number : 942404-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131372

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131372
Availability details for
Quantity
  • Availability
  • Add to Cart