ENA402266916 Enamine

tert-butyl N-{[3-(4-phenylphenyl)-4,5-dihydro-1,2-oxazol-5-yl]methyl}carbamate

  • MDL number MFCD25371706
  • Cas Number : 1461705-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131378
Availability details for
Quantity
  • Availability
  • Add to Cart