ENA402266922 Enamine

methyl 1-(3-fluorophenyl)-5-propyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD25371717
  • Cas Number : 1461706-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131449
Availability details for
Quantity
  • Availability
  • Add to Cart